APOH99994626
Apollo Scientific Ltd
1,2,4,5-Tetrafluoro-3-iodo-6-methoxybenzene
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
306
Pricing and availability is not currently available. Contact Technical Service
Iupac Name
1,2,4,5-tetrafluoro-3-iodo-6-methoxybenzene
Inchi Code
1S/C7H3F4IO/c1-13-7-4(10)2(8)6(12)3(9)5(7)11/h1H3
InChI key
ZMUUZYWIZVBAAZ-UHFFFAOYSA-N
Country of Origin
GB
Product Link
https://store.apolloscientific.co.uk/product/1245-tetrafluoro-3-iodo-6-methoxybenzene
MSDS
https://store.apolloscientific.co.uk/product/1245-tetrafluoro-3-iodo-6-methoxybenzene
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service