Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

725420

Sigma-Aldrich

1-Boc-2-(4-nitrobenzenesulfonyl)hydrazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15N3O6S
CAS Number:
Molecular Weight:
317.32
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

142-145 °C

functional group

amine
hydrazide
nitro

SMILES string

CC(C)(C)OC(=O)NNS(=O)(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C11H15N3O6S/c1-11(2,3)20-10(15)12-13-21(18,19)9-6-4-8(5-7-9)14(16)17/h4-7,13H,1-3H3,(H,12,15)

InChI key

NOJCYFMYHGQHAM-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Sens. 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service