Skip to Content
MilliporeSigma
All Photos(1)

Documents

469017

Sigma-Aldrich

6H-Dibenz[c,e][1,2]oxaphosphorin-6-oxide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9O2P
CAS Number:
Molecular Weight:
216.17
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

mp

118-121 °C (lit.)

SMILES string

O=[PH]1Oc2ccccc2-c3ccccc13

Application

Used to prepare 6-hydroxyalkyl/aryl substituted derivatives.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Heterocyclic Chemistry, 27, 845-845 (1990)
Amy Godfrey et al.
Aquatic toxicology (Amsterdam, Netherlands), 193, 228-235 (2017-11-05)
Endocrine disrupting chemicals (EDCs) can alter thyroid function and adversely affect growth and development. Halogenated compounds, such as perfluorinated chemicals commonly used in food packaging, and brominated flame retardants used in a broad range of products from clothing to electronics
Boris V Krivoshiev et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(8), 2124-2132 (2015-09-08)
Flame retardants are ubiquitously used chemicals that have been shown to contaminate environments. Toxicological data is largely limited, with little insight into their molecular modes of action that may give rise to their toxic phenotypes. Such insight would aid more

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service