Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

368210

Sigma-Aldrich

Ethyl 2-(trimethylsilylmethyl)acrylate

97%

Synonym(s):

2-(Trimethylsilylmethyl)acrylic acid ethyl ester, Ethyl 2-(trimethylsilylmethyl)propenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C[CH2Si(CH3)3]CO2C2H5
CAS Number:
Molecular Weight:
186.32
Beilstein/REAXYS Number:
4658894
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n20/D 1.438 (lit.)

bp

70-72 °C/10 mmHg (lit.)

density

0.897 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C(=C)C[Si](C)(C)C

InChI

1S/C9H18O2Si/c1-6-11-9(10)8(2)7-12(3,4)5/h2,6-7H2,1,3-5H3

InChI key

HSEYTIDQGJXPIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Carbohydrate Derived ?-Methylene-?-lactones by Diasterecseldctive, Low Temperature Reformatsky-Type Reactions
Csuk R, et al.
Journal of Carbohydrate Chemistry, 5(3), 459-467 (1986)
Synthesis of Carbohydrate Derived a-Methylene-?-lactones by Diasterecseldctive, Low Temperature Reformatsky-Type Reactions.
Csuk R, et al.
Journal of Carbohydrate Chemistry, 5(3), 459- 467 (1986)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service