Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

103926

Sigma-Aldrich

Dimethoxy(methyl)phenylsilane

95%

Synonym(s):

Methyl-phenyl-dimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O2Si
CAS Number:
Molecular Weight:
182.29
Beilstein/REAXYS Number:
1866944
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

95%

bp

199 °C (lit.)

density

1.005 g/mL at 20 °C (lit.)

SMILES string

CO[Si](C)(OC)c1ccccc1

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service