Skip to Content
MilliporeSigma
All Photos(1)

Documents

M8814

Sigma-Aldrich

Myristoyl-Phe-Ala-Arg-Lys-Gly-Ala-Leu-Arg-Gln

>95% (HPLC), solid

Synonym(s):

myr-PKC[19-27]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H105N17O12
CAS Number:
Molecular Weight:
1256.58
MDL number:
UNSPSC Code:
12352202

assay

>95% (HPLC)

form

solid

storage temp.

−20°C

Biochem/physiol Actions

Selective cell-permeable inhibitor of PKC in intact cells.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service