Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C6H5O(CH2)4Cl
CAS Number:
Molecular Weight:
184.66
UNSPSC Code:
12352103
EC Number:
220-174-3
MDL number:
assay
99%
refractive index
n
bp
147 °C/12 mmHg (lit.)
density
1.068 g/mL at 25 °C (lit.)
SMILES string
ClCCCCOc1ccccc1
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Storage Class
13 - Non Combustible Solids
WGK
WGK 3
Flash Point (°F)
>230.0 °F - closed cup
Flash Point (°C)
> 110 °C - closed cup
PPE (personal protective equipment)
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

