Skip to Content
MilliporeSigma

565776

5-Fluoro-2-propoxyphenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H12BFO3
CAS Number:
Molecular Weight:
198.00
UNSPSC Code:
12352103
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

mp

86-91 °C (lit.)

SMILES string

CCCOc1ccc(F)cc1B(O)O

InChI

1S/C9H12BFO3/c1-2-5-14-9-4-3-7(11)6-8(9)10(12)13/h3-4,6,12-13H,2,5H2,1H3

InChI key

TVQRNJJJJCUOLL-UHFFFAOYSA-N

Other Notes

Contains varying amounts of anhydride


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library