Skip to Content
MilliporeSigma

540595

(S)-(−)-3,3′-Dibromo-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-2,2′-naphthalenediol

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20Br2O2
CAS Number:
Molecular Weight:
452.18
PubChem Substance ID:
UNSPSC Code:
12352101
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

97%

optical activity

[α]20/D −53°, c = 1 in THF

mp

145.5-148.5 °C (lit.)

functional group

bromo

InChI

1S/C20H20Br2O2/c21-15-9-11-5-1-3-7-13(11)17(19(15)23)18-14-8-4-2-6-12(14)10-16(22)20(18)24/h9-10,23-24H,1-8H2

InChI key

WDRTXCNGVVLRSZ-UHFFFAOYSA-N

General description

(S)-3,3′-Dibromo-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-2,2′-naphthalenediol can be used in the synthesis of chiral bidentate hybrid ligands for asymmetric hydrogenation and hydroformylation reactions.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library