Skip to Content
MilliporeSigma

499765

2,5-Dimethoxy-3-nitrobenzoic acid

99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
O2NC6H2(OCH3)2CO2H
CAS Number:
Molecular Weight:
227.17
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

mp

183-187 °C (lit.)

SMILES string

COc1cc(C(O)=O)c(OC)c(c1)[N+]([O-])=O

InChI

1S/C9H9NO6/c1-15-5-3-6(9(11)12)8(16-2)7(4-5)10(13)14/h3-4H,1-2H3,(H,11,12)

InChI key

QCJROOYLFVYZEP-UHFFFAOYSA-N

General description

2,5-Dimethoxy-3-nitrobenzoic acid is a substituted nitrobenzoic acid.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library