Skip to Content
MilliporeSigma

424153

3-Methyl-5-(trifluoromethyl)pyrazole

99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5F3N2
CAS Number:
Molecular Weight:
150.10
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
99%
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

mp

88-90 °C (lit.)

SMILES string

Cc1cc(n[nH]1)C(F)(F)F

InChI

1S/C5H5F3N2/c1-3-2-4(10-9-3)5(6,7)8/h2H,1H3,(H,9,10)

InChI key

DLCHCAYDSKIFIN-UHFFFAOYSA-N

General description

3-Methyl-5-(trifluoromethyl)pyrazole is a pyrazole derivative. It has been reported to be synthesized from 3-methyl-5-trifluoromethyl-1H-pyrazole.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library