Skip to Content
MilliporeSigma

253286

4-Isothiocyanato-TEMPO

Synonym(s):

4-Isothiocyanato-2,2,6,6-tetramethyl-1-piperidinyloxy, free radical

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H18N2OS
CAS Number:
Molecular Weight:
214.33
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

CC1(C)CC(CC(C)(C)N1[O])N=C=S

Features and Benefits

Spin label.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library