Skip to Content
MilliporeSigma

150207

tert-Butyl 2,4,5-trichlorophenyl carbonate

99%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)3COCO2C6H2Cl3
CAS Number:
Molecular Weight:
297.56
UNSPSC Code:
12352100
PubChem Substance ID:
EC Number:
241-039-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

99%

mp

66-68 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)Oc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C11H11Cl3O3/c1-11(2,3)17-10(15)16-9-5-7(13)6(12)4-8(9)14/h4-5H,1-3H3

InChI key

LJFPGBIHDPZHIN-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library