Merck
Photoisomerization of Protonated Azobenzenes in the Gas Phase.
The journal of physical chemistry. A (2017-08-05)
Michael S Scholz, James N Bull, Neville J A Coughlan, Eduardo Carrascosa, Brian D Adamson, Evan J Bieske
ABSTRACT
Because of their high photoisomerization efficiencies, azobenzenes and their functionalized derivatives are used in a broad range of molecular photoswitches. Here, the photochemical properties of the trans isomers of protonated azobenzene (ABH
Sigma-Aldrich
Azobenzene, 98%