Merck
  • High frequency of diabetes and impaired fasting glucose in patients with glucose-6-phosphate dehydrogenase deficiency in the Western brazilian Amazon.
High frequency of diabetes and impaired fasting glucose in patients with glucose-6-phosphate dehydrogenase deficiency in the Western brazilian Amazon.
The American journal of tropical medicine and hygiene (2014-05-29)
Marli S Santana, Wuelton M Monteiro, Mônica R F Costa, Vanderson S Sampaio, Marcelo A M Brito, Marcus V G Lacerda, Maria G C Alecrim
ABSTRACT
Glucose-6-phosphate dehydrogenase (G6PD) deficiency is one of the most common human genetic abnormalities, and it has a significant prevalence in the male population (X chromosome linked). The purpose of this study was to estimate the frequency of impaired fasting glucose and diabetes among G6PD-deficient persons in Manaus, Brazil, an area in the Western Brazilian Amazon to which malaria is endemic. Glucose-6-phosphate dehydrogenase-deficient males had more impaired fasting glucose and diabetes. This feature could be used as a screening tool for G6PD-deficient persons who are unable to use primaquine for the radical cure of Plasmodium vivax malaria.
Product Description
Primaquine diphosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Primaquine bisphosphate, 98%
USP
Primaquine phosphate, United States Pharmacopeia (USP) Reference Standard