Merck
Effects of 4-acetylpyridine on stress ulcer formation in mice.
Neuroscience letters (1988-02-15)
A Hall, R Bose, G Glavin, C Pinsky
ABSTRACT
4-Acetylpyridine, earlier reported by us to be an anticonvulsant, offers long-lasting protection after a single administration against hypothermic restraint stress-induced gastric ulceration in mice. Electroshock convulsions, marginally but not significantly protective against such ulcers themselves, when coupled with 4-acetylpyridine administration fully prevented gastric ulcers from occurring in this murine model of experimentally induced stress.
Sigma-Aldrich
4-Acetylpyridine, 97%