Merck
  • Synthesis, crystal structures, and biological evaluation of manganese(II) and nickel(II) complexes of 4-cyclohexyl-1-(1-(pyrazin-2-yl)ethylidene)thiosemicarbazide.
Synthesis, crystal structures, and biological evaluation of manganese(II) and nickel(II) complexes of 4-cyclohexyl-1-(1-(pyrazin-2-yl)ethylidene)thiosemicarbazide.
European journal of medicinal chemistry (2011-07-26)
Ming Xue Li, Li Zhi Zhang, Dong Zhang, Bian Sheng Ji, Jun Wei Zhao
ABSTRACT
4-Cyclohexyl-1-(1-(pyrazin-2-yl)ethylidene)thiosemicarbazide (HL) and its transition metal complexes formulated as [Mn(L)(2)] (1) and [Ni(L)(2)] (2) have been prepared in 55-75% yield and characterized by elemental analysis, IR, MS, NMR and single-crystal X-ray diffraction studies. Biological activities of the synthesized compounds have been evaluated against selected Gram positive bacteria Bacillus subtilis, Gram negative bacteria Pseudomonas aeruginosa and the K562 leukemia cell line, respectively. The cytotoxicity data suggest that these compounds may be endowed with important biological properties, especially the nickel complex 2 with MIC = 31.2 μg/mL and IC(50) = 0.53 μM, respectively. Effect of the free ligand and its two complexes on Mitochondria membrane potential (MMP) and PI-associated fluorescence intensity as well as their effect on cell apoptosis in K562 leukemia cell line was also studied. The tested compounds may exert their cytotoxicity activity via induced loss of MMP.
Sigma-Aldrich
Thiosemicarbazide, 99%
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%