Merck
  • Analysis of paraquat, diquat and two diquat metabolites in biological materials by high-performance liquid chromatography.
Analysis of paraquat, diquat and two diquat metabolites in biological materials by high-performance liquid chromatography.
Legal medicine (Tokyo, Japan) (2003-08-26)
Chiaki Fuke, Tomonori Arao, Yasumasa Morinaga, Hajime Takaesu, Kiyoshi Ameno, Tetsuji Miyazaki
ABSTRACT
The determination of paraquat, diquat and two metabolites of diquat in biological materials was developed using high-performance liquid chromatography combined with UV and fluorescence detectors. Paraquat, diquat and internal standards (ethyl viologen and o-acetamidophenol) were detected by the UV detector. Diquat-monopyridone and diquat-dipyridone were monitored by the fluorescence detector. Paraquat, diquat, diquat-monopyridone and ethyl viologen were effectively extracted from blood, liver and brain, using a Sep-Pak C(18) cartridge. Diquat-dipyridone and o-acetamidophenol were extracted with methanol. Paraquat and diquat at a concentration range of 0.1-10 microg/ml (or g), and diquat-monopyridone and diquat-dipyridone at a concentration range of 0.01-1 microg/ml (or g) in biological material were determined with high accuracy and precision. The detection limits of paraquat, diquat, diquat-monopyridone and diquat-dipyridone were 1, 1, 0.02 and 0.02 ng, respectively, as an injection amount. This method was applied for toxicological examination of a case of suspected paraquat and diquat intoxication. The analysis of the metabolites of diquat was helpful for the estimation of the elapsed time from ingestion to death.
Product Description
Supelco
Diquat metabolite Dipyridone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Diquat metabolite Monopyridone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland