
Paclitaxel impurity K

Synonym(s):
7-O-(Triethylsilanyl)paclitaxel, 7-O-(Triethylsilyl)taxol
Empirical Formula (Hill Notation):
C53H65NO14Si
CAS Number:
Molecular Weight:
968.17
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing