![Mycophenolate mofetil for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/300/411/d9aae4f8-9d91-4257-bdc8-6289ff6ee694/640/d9aae4f8-9d91-4257-bdc8-6289ff6ee694.png)
Mycophenolate mofetil for peak identification
![Mycophenolate mofetil for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/300/411/d9aae4f8-9d91-4257-bdc8-6289ff6ee694/640/d9aae4f8-9d91-4257-bdc8-6289ff6ee694.png)
Synonym(s):
Mycophenolate mofetil, (4E)-6-(1,3-Dihydro-4-hydroxy-6-methoxy-7-methyl-3-oxo-5-isobenzofuranyl)-4-methyl-4-hexenoic acid 2-(4-morpholinyl)ethyl ester, RS 61443, TM-MMF
Empirical Formula (Hill Notation):
C23H31NO7
CAS Number:
Molecular Weight:
433.49
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing