
D-Camphor Impurity A, a-pinene

Synonym(s):
α-Pinene, (±)-2-Pinene, 2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing