
Anagrelide Related Compound C

Synonym(s):
Ethyl 2-amino-5,6-dichloro-3(4H)-quinazolineacetate monohydrobromide, Ethyl 2-(5,6-dichloro-2-imino-1,2-dihydroquinazolin-3(4H)-yl)acetate hydrobromide
Empirical Formula (Hill Notation):
C12H13Cl2N3O2 · HBr
CAS Number:
Molecular Weight:
383.07
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2