HPLC Analysis of Glycan and Isomers on Proteomix® SAX-NP5, High Resolution Separation
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase
(A) 2.5% (v/v) acetic acid, 0.5% TEA in water; 0.5% acetic acid in acetonitrile
gradient
0-100% B in 60 min
flow rate
0.3 mL/min
detector
Fluorescence, Ex/Em=360/425 nm
sample
2-AA (anthranilic acid) labeled N-linked oligosaccharide profiling of an IgG1 sample
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies