Merck
  • Endothelial damage in major depression patients is modulated by SSRI treatment, as demonstrated by circulating biomarkers and an in vitro cell model.
Endothelial damage in major depression patients is modulated by SSRI treatment, as demonstrated by circulating biomarkers and an in vitro cell model.
Translational psychiatry (2016-09-07)
I Lopez-Vilchez, M Diaz-Ricart, V Navarro, S Torramade, J Zamorano-Leon, A Lopez-Farre, A M Galan, C Gasto, G Escolar
ABSTRACT
There is a link between depression, cardiovascular events and inflammation. We have explored this connection through endothelial dysfunction, using in vivo and in vitro approaches. We evaluated circulating biomarkers of endothelial dysfunction in patients with major depression at their diagnosis (MD-0) and during antidepressant treatment with the selective serotonin reuptake inhibitor escitalopram, for 8 and 24 weeks (MD-8 and MD-24). Results were always compared with matched healthy controls (CON). We measured in vivo circulating endothelial cells (CECs) and endothelial progenitor cells (EPCs) in blood samples, and assessed plasma levels of soluble von Willebrand factor (VWF) and vascular cell adhesion molecule-1 (VCAM-1). CEC counts, soluble VWF and VCAM-1 were statistically elevated in MD-0 (P<0.01 versus CON) and gradually decreased during treatment. Conversely, EPC levels were lower in MD-0, tending to increase throughout treatment. In vitro studies were performed in human endothelial cells cultured in the presence of sera from each study group. Elevated expression of the inflammation marker intercellular adhesion molecule-1 and oxidative stress, with lower presence of endothelial nitric oxide synthase and higher reactive oxygen species production, were found in cells exposed to MD-0 sera (P<0.05 versus CON). These results were normalized in cells exposed to MD-24 sera. Thrombogenicity of extracellular matrices generated by these cells, measured as expression of VWF, tissue factor and platelet reactivity, showed non-significant differences. We provide a model of cultured endothelial cells reproducing endothelial dysfunction in naive patients with major depression, demonstrating endothelial damage and inflammation at diagnosis, and recovering with selective serotonin reuptake inhibitor treatment for 24 weeks.
Sigma-Aldrich
Anti-ICAM-1 Antibody, clone P2A4, clone P2A4, Chemicon®, from mouse
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)