Merck
  • Preclinical evaluation of engineered oncolytic herpes simplex virus for the treatment of pediatric solid tumors.
Preclinical evaluation of engineered oncolytic herpes simplex virus for the treatment of pediatric solid tumors.
PloS one (2014-02-06)
Michael L Megison, Lauren A Gillory, Jerry E Stewart, Hugh C Nabers, Elizabeth Mroczek-Musulman, Alicia M Waters, Jennifer M Coleman, Virginia Kelly, James M Markert, G Yancey Gillespie, Gregory K Friedman, Elizabeth A Beierle
ABSTRACT
Recently, investigators showed that mice with syngeneic murine gliomas that were treated with a neuroattenuated oncolytic herpes simplex virus-1 (oHSV), M002, had a significant increase in survival. M002 has deletions in both copies of the γ134.5 gene, enabling replication in tumor cells but precluding infection of normal cells. Previous studies have shown antitumor effects of other oHSV against a number of adult tumors including hepatocellular carcinoma and renal cell carcinoma. The purpose of the current study was to investigate the oncolytic potential of M002 against difficult to treat pediatric liver and kidney tumors. We showed that the oHSV, M002, infected, replicated, and decreased cell survival in hepatoblastoma, malignant rhabdoid kidney tumor, and renal sarcoma cell lines. In addition, we showed that in murine xenografts, treatment with M002 significantly increased survival and decreased tumor growth. Finally, these studies showed that the primary entry protein for oHSV, CD111 (nectin-1) was present in human hepatoblastoma and malignant rhabdoid kidney tumor specimens. We concluded that M002 effectively targeted these rare aggressive tumor types and that M002 may have potential for use in children with unresponsive or relapsed pediatric solid tumors.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
May-Grünwald Stain, Suitable for the differential staining of cellular elements of blood