Merck
  • Cilostazol versus aspirin for secondary prevention of vascular events after stroke of arterial origin.
Cilostazol versus aspirin for secondary prevention of vascular events after stroke of arterial origin.
The Cochrane database of systematic reviews (2011-01-21)
Ayeesha K Kamal, Imama Naqvi, Muhammad R Husain, Bhojo A Khealani
ABSTRACT
Aspirin is widely used for secondary prevention after stroke. Cilostazol has shown promise as an alternative to aspirin in Asian people with stroke. To determine the relative effectiveness and safety of cilostazol compared directly with aspirin in the prevention of stroke and other serious vascular events in patients at high vascular risk for subsequent stroke, those with previous transient ischaemic attack (TIA) or ischaemic stroke of arterial origin. We searched the Cochrane Stroke Group Trials Register (last searched September 2010), the Cochrane Central Register of Controlled Trials (CENTRAL) (The Cochrane Library 2009, Issue 4), MEDLINE (1950 to May 2010) and EMBASE (1980 to May 2010). In an effort to identify further published, ongoing and unpublished studies we searched journals, conference proceedings and ongoing trial registers, scanned reference lists from relevant studies and contacted trialists and Otsuka Pharmaceutical Co Ltd. We selected all randomised controlled trials (RCTs) comparing cilostazol with aspirin where participants were treated for at least one month and followed systematically for development of vascular events. Data extracted from eligible studies included: (1) a composite outcome of vascular events (stroke, myocardial infarction or vascular death) during follow up (primary outcome); (2) separate outcomes of stroke (ischaemic or haemorrhagic, fatal or non-fatal), myocardial infarction (MI) (fatal or non-fatal), vascular death and death from all causes; and (3) main outcomes of safety including any intracranial, extracranial or gastrointestinal (GI) haemorrhage and other outcomes during treatment follow up (secondary outcomes). We computed an estimate of treatment effect and performed a test for heterogeneity between trials. We analysed data on an intention-to-treat basis and assessed bias for all included studies. We included two RCTs with 3477 Asian participants. Compared with aspirin, cilostazol was associated with a significantly lower risk of composite outcome of vascular events (6.77% versus 9.39%, risk ratio (RR) 0.72, 95% confidence interval (CI) 0.57 to 0.91), and lower risk of haemorrhagic stroke (0.53% versus 2.01%, RR 0.26, 95% CI 0.13 to 0.55). In terms of outcome of safety compared with aspirin, cilostazol was significantly associated with minor adverse effects (8.22% versus 4.95%, RR 1.66, 95% CI 1.51 to 1.83). Cilostazol is more effective than aspirin in the prevention of vascular events secondary to stroke. Cilostazol has more minor adverse effects, although there is evidence of fewer bleeds.
Product Description
USP
Cilostazol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cilostazol, ≥98% (HPLC), powder