Merck
Effect of pencycuron on microbial parameters of waterlogged soil.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2006-11-09)
R Pal, K Chakrabarti, A Chakraborty, A Chowdhury
ABSTRACT
A laboratory study was conducted to monitor the effect of pencycuron [1-(4-chlorobenzyl)-1-cyclopentyl-3-phenylurea] on microbial parameters of alluvial (AL) soil (Typic udifluvent) and coastal saline (CS) soil (Typic endoaquept) under waterlogged condition. Pencycuron at field rate (FR), 2FR and 10FR affected the microbial biomass C (MBC), soil ergosterol content and fluorescein diacetate hydrolyzing activity (FDHA) differentially. The DCM amendment did not seem to have any counteractive effect on the toxicity of pencycuron on the microbial variables. The change in microbial metabolic quotient (qCO2) and microbial respiration quotient (QR), indicated pencycuron induced disturbance at 10FR. Present study revealed that the metabolically activated microbial population was more suppressed compared to the dormant population.
Product Description
Supelco
Pencycuron, PESTANAL®, analytical standard