Merck
  • Nonheme iron-oxo and -superoxo reactivities: O2 binding and spin inversion probability matter.
Nonheme iron-oxo and -superoxo reactivities: O2 binding and spin inversion probability matter.
Chemical communications (Cambridge, England) (2012-01-19)
Kyung-Bin Cho, Hui Chen, Deepa Janardanan, Sam P de Visser, Sason Shaik, Wonwoo Nam
ABSTRACT
DFT calculated barriers for C-H activation of 1,4-cyclohexadiene by nonheme iron(IV)-oxo and iron(III)-superoxo species show that the experimental trends can be explained if the spin inversion probability of the TMC iron(IV)-oxo is assumed to be poor. Also, the TMC iron(III)-superoxo reaction proceeds with an endothermic O(2)-binding energy followed by an intrinsically reactive quintet state.
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
1,4-Cyclohexadiene, 97%