Merck
  • DNA single strand breaks in peripheral lymphocytes associated with urinary thiodiglycolic acid levels in polyvinyl chloride workers.
DNA single strand breaks in peripheral lymphocytes associated with urinary thiodiglycolic acid levels in polyvinyl chloride workers.
Mutation research (2004-07-09)
Yu-Chen Lei, Huei-Ting Yang, Yee-Chung Ma, Ming-Feng Huang, Wushou P Chang, Tsun-Jen Cheng
ABSTRACT
The association between vinyl chloride monomer (VCM) exposure and DNA damage has been established. However, the relationship between individual exposure and DNA single strand breaks was limited. Since environmental monitoring may not reflect the actual exposure, a useful marker of exposure is needed to assess the individual exposure. In our previous study, we have found a high correlation between air VCM level and urinary thiodiglycolic acid (TdGA) at the commencement of the next shift. Here, we further used comet assay to evaluate the relationship between urinary TdGA levels and DNA single strand breaks in polyvinyl chloride monomer (PVC) workers. Urinary TdGA levels (n=26) at the commencement of the following shift were analyzed. Ten of the 26 workers also had personal air sampling for air VCM exposure. Questionnaires were administered to obtain epidemiological information including detailed history of occupation and lifestyles. Workers experiencing air VCM level greater than 5 ppm had higher tail moment and tail intensity (%) than those experiencing VCM exposure between 1 and 5, or <1 ppm, respectively (P < 0.05). The results also revealed that level of DNA single strand breaks, including tail moment and tail intensity, were increased with urinary TdGA level. The dose-response relationship of urinary TdGA level and DNA single strand breaks was particularly significant among the workers with 4 mg/g Cr of urinary TdGA level, which is equivalent to 5 ppm air VCM level. We concluded that air VCM exposure greater than 5 ppm could induce DNA damage. Further sensitive assay should be developed for the diction of DNA damage when air VCM exposure below 5 ppm.
Sigma-Aldrich
2,2′-Thiodiacetic acid, 98%