Merck
  • Pharmacological interventions in the newborn piglet in the first 24 h after hypoxia-ischemia. A hemodynamic and electrophysiological perspective.
Pharmacological interventions in the newborn piglet in the first 24 h after hypoxia-ischemia. A hemodynamic and electrophysiological perspective.
Experimental brain research (2002-11-01)
Cacha Peeters-Scholte, Evelyn van den Tweel, Tomoaki Ioroi, Ilka Post, Kees Braun, Wouter Veldhuis, Klaas Nicolay, Floris Groenendaal, Frank van Bel
ABSTRACT
The purpose of this study was to investigate whether combined inhibition of neuronal and inducible nitric oxide synthase (NOS) by 2-iminobiotin, free radical scavenging by allopurinol, and non-protein-bound iron chelation with deferoxamine improved cerebral oxygenation, electrocortical brain activity, and brain energy status during the first 24 h after hypoxia-ischemia (HI) in the newborn piglet. Forty-three newborn piglets were subjected to 1 h of severe HI by occluding both carotid arteries and phosphorous magnetic resonance spectroscopy ((31)P-MRS)-guided hypoxia, whereas five served as sham-operated controls. Upon reperfusion, piglets received vehicle (n=12), 2-iminobiotin (n=11), allopurinol (n=10), or deferoxamine (n=10). Cerebral oxygenation was recorded with near-infrared spectrophotometry (NIRS), electrocortical brain activity was assessed with amplitude-integrated EEG (aEEG), and cerebral energy status with (31)P-MRS. The oxygenated hemoglobin (HbO(2)) and total hemoglobin (tHb) were significantly increased in vehicle-treated piglets compared with 2-iminobiotin-treated and deferoxamine-treated piglets. No change in deoxygenated Hb (HHb) was demonstrated over time. The aEEG was significantly preserved in 2-iminobiotin- and deferoxamine-treated piglets compared with vehicle-treated piglets. Allopurinol treatment was not as effective as 2-iminobiotin treatment after HI. Phosphocreatine/inorganic phosphate ratios (PCr/P(i)) were significantly decreased for vehicle-treated piglets at 24 h post-HI, whereas 2-iminobiotin, allopurinol, and deferoxamine prevented the development of secondary energy failure. We speculate that the beneficial effects, especially of 2-iminobiotin, but also of deferoxamine, are due to reduced peroxynitrite-mediated oxidation.
Sigma-Aldrich
2-Iminobiotin, ≥98% (TLC)