Merck
  • Bone Regeneration and Angiogenesis by Co-transplantation of Angiotensin II-Pretreated Mesenchymal Stem Cells and Endothelial Cells in Early Steroid-Induced Osteonecrosis of the Femoral Head.
Bone Regeneration and Angiogenesis by Co-transplantation of Angiotensin II-Pretreated Mesenchymal Stem Cells and Endothelial Cells in Early Steroid-Induced Osteonecrosis of the Femoral Head.
Cell transplantation (2022-03-23)
Jingjing Zhao, Wei He, Hongqing Zheng, Rui Zhang, Hao Yang
ABSTRACT
Mesenchymal stem cells (MSCs) have been shown to exert a positive impact on osteonecrosis of the femoral head (ONFH) in preclinical experiments and clinical trials. After the femoral head suffers avascular necrosis, the transplanted MSCs undergo a great deal of stress-induced apoptosis and senescence in this microenvironment. So, survival and differentiation of MSCs in osteonecrotic areas are especially important in ONFH. Although MSCs and endothelial cells (ECs) co-culture enhancing proliferation and osteogenic differentiation of MSCs and form more mature vasculature in vivo, it remains unknown whether the co-culture cells are able to repair ONFH. In this study, we explored the roles and mechanisms of co-transplantation of angiotensin II (Ang II)-MSCs and ECs in repairing early ONFH. In vitro, when MSCs and ECs were co-cultured in a ratio of 5:1, both types of cells managed to proliferate and induce both osteogenesis and angiogenesis. Then, we established a rabbit model of steroid-induced ONFH and co-transplantation of Ang II-MSCs and ECs through the tunnel of core decompression. Four weeks later, histological and Western blot analyses revealed that ONFH treated with Ang II-MSCs and ECs may promote ossification and revascularization by increasing the expression of collagen type I, runt-related transcription factor 2, osteocalcin, and vascular endothelial growth factor in the femoral head. Our data suggest that co-transplantation of Ang II-MSCs and ECs was able to rescue the early steroid-induced ONFH via promoting osteogenesis and angiogenesis, which may be regarded as a novel therapy for the treatment of ONFH in a clinical setting.
Sigma-Aldrich
Anti-RUNX2 (AB1) antibody produced in rabbit, IgG fraction of antiserum