Merck
  • A parathyroid hormone-related protein implicated in malignant hypercalcemia: cloning and expression.
A parathyroid hormone-related protein implicated in malignant hypercalcemia: cloning and expression.
Science (New York, N.Y.) (1987-08-21)
L J Suva, G A Winslow, R E Wettenhall, R G Hammonds, J M Moseley, H Diefenbach-Jagger, C P Rodda, B E Kemp, H Rodriguez, E Y Chen
ABSTRACT
Humoral hypercalcemia of malignancy is a common complication of lung and certain other cancers. The hypercalcemia results from the actions of tumor factors on bone and kidney. We report here the isolation of full-length complementary DNA clones of a putative hypercalcemia factor, and the expression from the cloned DNA of the active protein in mammalian cells. The clones encode a prepro peptide of 36 amino acids and a mature protein of 141 amino acids that has significant homology with parathyroid hormone in the amino-terminal region. This previously unrecognized hormone may be important in normal as well as abnormal calcium metabolism.
Sigma-Aldrich
PTHrP human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
Hypercalcemia of malignancy factor 1-40 human, ≥95% (HPLC)