Merck
  • Evidence for endotoxin contamination in plastic Na+-heparin blood collection tube lots.
Evidence for endotoxin contamination in plastic Na+-heparin blood collection tube lots.
Clinical chemistry (2010-07-29)
Kathryn J Newhall, Geoffrey S Diemer, Natalia Leshinsky, Keith Kerkof, Hilary T Chute, Chris B Russell, William Rees, Andrew A Welcher, Scott D Patterson, Gary D Means
ABSTRACT
Biomarker assays are often conducted on whole blood samples in the course of drug development studies. Because bacterial lipopolysaccharide (LPS) (endotoxin) contamination is known to cause spontaneous cytokine production by monocytes, contamination of blood collection tubes may interfere with biomarker assay results. Whole blood from healthy donors was collected into plastic or glass sodium (Na(+))-heparin Vacutainer() blood collection tubes and heparinized syringes. Samples were analyzed for phosphoprotein response, cytokine production, and RNA expression. Tubes were tested for endotoxin contamination by use of the limulus amoebocyte lysate assay. Results of phospho-flow cytometry, branched DNA (bDNA), and ELISA assays indicated that a specific lot (#5339582) of plastic Na(+)-heparin Vacutainer tubes was highly contaminated with an endotoxinlike substance, and contamination was confirmed by the limulus amoebocyte lysate assay. Analysis of multiple-analyte panels revealed that analytes whose changed expression was predictive of LPS stimulation were increased when whole blood was incubated in contaminated tubes for 6 or 18 h. Two additional lots of plastic tubes tested had detectable amounts of endotoxin sufficient to strongly alter phospho-flow cytometry analyses, as determined by the fold change in phosphorylation of p38 mitogen-activated protein kinase in response to tumor necrosis factor alpha and LPS. In contrast, 3 lots of glass tubes had substantially lower levels of spontaneous blood activation. Endotoxin contamination associated with tubes from 3 lots of a particular type of plastic Na(+)-heparin Vacutainer tube dramatically affected biomarker assay measurements. Prescreening these tubes is suggested before their use in clinical sample analysis.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, γ-irradiated, BioXtra, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Albumin, Bovine Serum, Fraction V, Fatty Acid-Poor, Endotoxin-Free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg