
Isofraxidin

Synonym(s):
6,8-Dimethoxyumbelliferone, 7-Hydroxy-6,8-dimethoxychromen-2-one, 7-Hydroxy-6,8-dimethoxycoumarin
Empirical Formula (Hill Notation):
C11H10O5
CAS Number:
Molecular Weight:
222.19
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing