
Hymecromone impurity A

Synonym(s):
Resorcinol, 1,3-Benzenediol
Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing