
4-(2-Hydroxy-3-isopropylaminopropoxy)phenylacetic acid

Synonym(s):
4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetic acid, Atenolol acid, Metoprolol acid
Empirical Formula (Hill Notation):
C14H21NO4
CAS Number:
Molecular Weight:
267.32
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing