
2-ethynylthiophene

Synonym(s):
2-Acetylenylthiophene
Empirical Formula (Hill Notation):
C6H4S
CAS Number:
Molecular Weight:
108.16
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing