Skip to Content
Merck
All Photos(1)

Key Documents

MABT1506

Sigma-Aldrich

Anti-Desmoglein 3 Antibody, clone 5H10

clone 5H10, from mouse

Synonym(s):

130 kDa pemphigus vulgaris antigen, PVA, Cadherin family member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5H10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DSG3(1830)

General description

Desmoglein-3 (UniProt: P32926; also known as 130 kDa pemphigus vulgaris antigen, PVA, Cadherin family member 6) is encoded by the DSG3 (also known as CDHF6) gene (Gene ID: 1830) in human. Desmogleins are protein substances located in and on the surface of keratinocytes. They are present in specialized cell-cell adhesion structures enriched in tissues that undergo substantial mechanical stress. These proteins have been shown to be a critical factor in cell-to-cell adhesion. Desmoglein-3 is a single-pass type I membrane protein that is a component of intercellular desmosome junctions. It is involved in the interaction of plaque proteins and intermediate filaments mediating cell-cell adhesion. It is synthesized with a signal peptide (aa 1-23) and a propeptide (aa 24-49) that are cleaved off in the mature form. It contains an extracellular domain (aa 50-615), a transmembrane domain (aa 616-640), and a cytoplasmic domain (aa 641-999). Desmoglein-3 has four cadherin domains and two desmoglein repeats. This antibody (clone 5H10) recognized the extracellular domain of desmoglein-3.

Specificity

Clone 5H10 detects Desmoglein-3 in human cells. It targets an epitope with in the extracellular domain.

Immunogen

Epitope: extracellular domain
Extracellular domain of Desmoglein 3

Application

Anti-Desmoglein 3, clone 5H10, Cat. No. MABT1506, is a mouse monoclonal antibody that detects Desmoglein-3 and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected Desmoglein 3 in Immunoprecipitation applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).

Flow Cytometry Analysis: A representative lot detected Desmoglein 3 in Flow Cytometry applications (Wan, H., et. al. (2007). Stem Cells. 25(5):1286-97).

Western Blotting Analysis: A representative lot detected Desmoglein 3 in Western Blotting applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).

Immunocytochemistry Analysis: A representative lot detected Desmoglein 3 in Immunocytochemistry applications (Wahl, J.K. 3rd, et. al. (2002). Hybrid Hybridomics. 21(1):37-44).

Immunofluorescence Analysis: A representative lot detected Desmoglein 3 in Immunofluorescnece applications (Wan, H., et. al. (2007). Stem Cells. 25(5):1286-97).
Research Category
Cell Structure

Quality

Evaluated by Western Blotting in HaCaT cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Desmoglein 3 in HaCaT cell lysate.

Target description

~125 kDa observed; 107.53 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service