HPLC Analysis of Ketoconazole Enantiomers on Astec® Cellulose DMP, No Additives
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 μm particles (51098AST)
mobile phase
[A] acetonitrile; (100, A)
flow rate
0.5 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in mobile phase