Merck
  • Chemical and enzyme-assisted syntheses of norbuprenorphine-3-β-D-glucuronide.
Chemical and enzyme-assisted syntheses of norbuprenorphine-3-β-D-glucuronide.
Bioconjugate chemistry (2011-03-26)
Jinda Fan, Sarah M Brown, Zhude Tu, Evan D Kharasch
ABSTRACT
Norbuprenorphine-3-β-d-glucuronide (nBPN-3-β-d-G, 1) is a major phase II metabolite of buprenorphine, a pharmaceutical used for the treatment of opioid addiction. The pharmacological activity of compound 1 is not clear because investigations have been limited by the lack of chemically pure, well characterized 1 in sufficient quantities for in vitro and in vivo experiments. This work describes two concise, new methods of synthesis of 1, a chemical and an enzyme-assisted synthesis. The chemical synthesis used a strategy based on a combination of Koenig-Knorr coupling and amino-silyl protection. The enzyme-assisted synthesis used dog liver to convert the substrate norbuprenorphine (nBPN, 2) to 1. Both methods provided 1, characterized by (1)H NMR and tandem mass spectrometry, with purity >96%. The fractional yield of the enzyme-assisted synthesis was greater than that of the chemical synthesis (67% vs 5.3%), but due to larger reaction volumes, the chemical synthesis afforded greater amounts of total 1.
Product Description
Supelco
Norbuprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norbuprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®