Merck
  • Nucleotide sequence of the Clostridium stercorarium xynB gene encoding an extremely thermostable xylanase, and characterization of the translated product.
Nucleotide sequence of the Clostridium stercorarium xynB gene encoding an extremely thermostable xylanase, and characterization of the translated product.
Bioscience, biotechnology, and biochemistry (1995-01-01)
M Fukumura, K Sakka, K Shimada, K Ohmiya
ABSTRACT
The nucleotides of the xynB gene of Clostridium stercorarium F-9 were sequenced. The structural gene consists of an open reading frame of 1161 bp encoding a xylanase (XynB) in family F of 387 amino acids with a molecular weight of 44,377. The molecular weight of the enzyme purified from a recombinant Escherichia coli was around 41,000, smaller than the predicted value, on SDS-polyacrylamide gel electrophoresis due to the lack of 32 amino acids at the N-terminus. Intact XynB with a molecular weight of around 43,000 was immunologically detected in the total cell proteins of a recombinant E. coli and C. stercorarium F-9. The purified XynB was active toward xylan, carboxymethylcellulose, p-nitrophenyl-beta-D-xylopyranoside and p-nitrophenyl-beta-D-cellobioside. The pH optimum was 7.0 and it was quite stable over the pH range of 5 to 12 at 4 degrees C. This enzyme was optimally active at 80 degrees C and retained about 50% of the original activity even after incubation at 100 degrees C for 10 min.
Sigma-Aldrich
4-Nitrophenyl β-D-cellobioside, ≥98% (TLC)