Merck
  • Characterization of the antioxidant activity of sugars and polyhydric alcohols in fish oil emulsions.
Characterization of the antioxidant activity of sugars and polyhydric alcohols in fish oil emulsions.
Journal of agricultural and food chemistry (2004-11-13)
Habibollah Faraji, Robert C Lindsay
ABSTRACT
Polyols have been incorporated into fish oil emulsions as a means for the inhibition of lipid oxidation and suppression of fishy flavor. However, the role of sugars and polyhydric alcohols as antioxidants has not been clearly established. Selected polyols were evaluated for their performance as antioxidants and modifiers of oxidation pathways in a model system. Oil/water (O/W) emulsions were prepared with freshly steam-deodorized menhaden oil. A layer of emulsion in aluminum pans held at 5 degrees C was exposed to 2550 lx fluorescent lights for 24 h before peroxide values and volatile flavor compounds were analyzed by GC headspace entrainment procedure. Antioxidant activity was confirmed for fructose, sucrose, raffinose, sorbitol, or mannitol when incorporated at 16% of the aqueous phase into model fish oil-in-water emulsions. Peroxide values were suppressed 10-18% in treated samples compared to control samples. Viscosity data did not exclude possible contributions from a restricted oxygen diffusion mechanism in the antioxidant activity, but revealed that emulsion viscosity did not govern fish oil oxidation rates. Combining polyols with phenolic antioxidants (alpha-tocopherol, BHT, or TBHQ) frequently diminished the antioxidant activity compared to that for individual phenolic antioxidants, which was interpreted as indicating that the H-donating activity of phenolic antioxidants was hindered by the H-bonding activity of polyols. A viscosity-based inhibition of the retroaldol conversion of (E,Z)-2,6-nonadienal to (Z)-4-heptenal with a high fructose concentration (67%) was attributed to a restriction of molecular mobility of reactants, but the conversion was only slightly inhibited by the concentration of fructose (16%) used in experimental emulsions. The data supported a hypothesis that either or both free radical scavenging and transition state metal chelation activities were provided by polyols in fish oil emulsions. Also, polyols retarded the water-requiring retroaldol decomposition of (E,Z)-2,6-nonadienal to (Z)-4-heptenal in the model systems and the reaction may be involved in some suppression of fishy flavors in emulsions.
Sigma-Aldrich
trans-2,cis-6-Nonadienal, mixture of isomers, ≥96%, stabilized, FCC, FG
Sigma-Aldrich
trans-2,cis-6-Nonadienal, 95%
Sigma-Aldrich
trans,trans-2,6-Nonadienal, ≥95%, stabilized, FG