Merck
  • Prevalence of metallo-beta-lactamase among Pseudomonas aeruginosa and Acinetobacter baumannii in a Korean university hospital and comparison of screening methods for detecting metallo-beta-lactamase.
Prevalence of metallo-beta-lactamase among Pseudomonas aeruginosa and Acinetobacter baumannii in a Korean university hospital and comparison of screening methods for detecting metallo-beta-lactamase.
Journal of microbiological methods (2003-07-05)
Eun-Jee Oh, Seungok Lee, Yeon-Joon Park, Jung Jun Park, Kanggyun Park, Sang-Il Kim, Moon Won Kang, Byung Kee Kim
ABSTRACT
To identify the metallo-beta-lactamases (MBLs) prevalent in Korea, a total of 130 clinical isolates of Pseudomonas aeruginosa and Acinetobacter baumannii (99 P. aeruginosa and 31 A. baumannii) with a reduced susceptibility to imipenem (IPM) and/or ceftazidime (CAZ) was subjected to PCR analyses with primers specific to bla(IMP-1), bla(VIM-1), and bla(VIM-2). In addition, inhibitor-potentiated disk diffusion methods (IPD) using two kinds of substrate-inhibitor combinations (ceftazidime-2-mercaptopropionic acid (2MPA) and imipenem-EDTA) were investigated. Thirty-three isolates (29 P. aeruginosa and 4 A. baumannii) carried bla(VIM-2) and two P. aeruginosa isolates harbored bla(IMP-1). The enterobacterial repetitive intergenic consensus PCR (ERIC-PCR) pattern revealed that many of the VIM-2-producing P. aeruginosa isolates were clonally related, whereas the A. baumannii isolates were diverse. The inhibitor-potentiated disk diffusion test using imipenem-EDTA was highly sensitive and specific for detecting the VIM-2 producer. These results suggest that VIM-2 is an important MBL in P. aeruginosa and A. baumannii in the Korean hospital of this study and that the IMP-1-producing P. aeruginosa has also emerged. Screening for MBLs and strict infection control for these isolates will contribute to prevent further spread of resistance.
Sigma-Aldrich
2-Mercaptopropionic acid, ≥95%, FG
Sigma-Aldrich
Thiolactic acid, 95%