Merck
  • Antisense to transforming growth factor-beta(1) facilitates the apoptosis of macrophages in rat vein grafts.
Antisense to transforming growth factor-beta(1) facilitates the apoptosis of macrophages in rat vein grafts.
Journal of vascular research (2008-03-21)
Nicholas S Heaton, Randal A Wolff, Rita L Malinowski, Debra A Hullett, John R Hoch
ABSTRACT
The success of peripheral vein grafts is limited by intimal hyperplasia. Transforming growth factor (TGF)-beta(1) has effects on cell proliferation, apoptosis and extracellular matrix synthesis. We have previously observed positive changes in vessel healing with antisense to TGF-beta(1). Adenovirus was used to transduce rat femoral artery vein grafts with antisense to TGF-beta(1) (Ad-AST) or the sequence encoding the bioactive form of TGF-beta(1) (Ad-BAT). Grafts were harvested at 1, 2, 4 and 12 weeks and formalin fixed for immunohistochemical studies of the cell markers proliferating cellular nuclear antigen (proliferation) and active caspase 3 (apoptosis). In situ DNA fragmentation assays were also performed to confirm active caspase 3 results. Ad-AST treatment significantly (p = 0.05) increased apoptosis of macrophages inside the internal elastic lamina. In addition, Ad-AST treatment significantly increased the cellularity of the graft at early time points and reduced it at later time points (p = 0.01). The low levels of TGF-beta(1) in Ad-AST treatment promote apoptosis of macrophages and provide an environment that is more conducive to the proliferation or infiltration of cells that contribute to healthy vessels.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - FITC antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Caldesmon (Smooth) antibody produced in mouse, clone hHCD, ascites fluid