Merck
  • The molecular landscape of neural differentiation in the developing Drosophila brain revealed by targeted scRNA-seq and multi-informatic analysis.
The molecular landscape of neural differentiation in the developing Drosophila brain revealed by targeted scRNA-seq and multi-informatic analysis.
Cell reports (2021-04-29)
Nigel S Michki, Ye Li, Kayvon Sanjasaz, Yimeng Zhao, Fred Y Shen, Logan A Walker, Wenjia Cao, Cheng-Yu Lee, Dawen Cai
ABSTRACT
The Drosophila type II neuroblast lineages present an attractive model to investigate the neurogenesis and differentiation process as they adapt to a process similar to that in the human outer subventricular zone. We perform targeted single-cell mRNA sequencing in third instar larval brains to study this process of the type II NB lineage. Combining prior knowledge, in silico analyses, and in situ validation, our multi-informatic investigation describes the molecular landscape from a single developmental snapshot. 17 markers are identified to differentiate distinct maturation stages. 30 markers are identified to specify the stem cell origin and/or cell division numbers of INPs, and at least 12 neuronal subtypes are identified. To foster future discoveries, we provide annotated tables of pairwise gene-gene correlation in single cells and MiCV, a web tool for interactively analyzing scRNA-seq datasets. Taken together, these resources advance our understanding of the neural differentiation process at the molecular level.
Sigma-Aldrich
Collagenase type I, The collagenase type I (from Clostridium histolyticum) is a crude collagenase preparation that can be used for the isolation of primary cells or for tissue dissociation by enzymatic means.
Pricing and availability is not currently available.