
Methylatropine bromide

Synonym(s):
Atropine methyl bromide
Empirical Formula (Hill Notation):
C18H26NO3 · Br
CAS Number:
Molecular Weight:
384.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing