
Mesotrione

Synonym(s):
2-(4-Mesyl-2-nitrobenzoyl)-1,3-cyclohexanedione
Empirical Formula (Hill Notation):
C14H13NO7S
CAS Number:
Molecular Weight:
339.32
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing