
Ingenol-3-angelate

Synonym(s):
(1aR,2S,5R,5aS,6S,8aS,9R,10aR)-5,5a-Dihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-1a,2,5,5a,6,9,10,10a-octahydro-1H-2,8a-methanocyclopenta[a]cyclpropa[e][10]annulen-6-yl (2Z)-2-methylbut-2-enoate, 3-Angeloylingenol, 3-Ingenyl angelate, I3A, Ingenol mebutate, PEP005
Empirical Formula (Hill Notation):
C25H34O6
CAS Number:
Molecular Weight:
430.53
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing