
Griseofulvin for LC assay and identification

Synonym(s):
Griseofulvin, (2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione
Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing