
Econazole for system suitability

Synonym(s):
Econazole nitrate, 1-[2-[(4-Chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole mononitrate, 1-{2,4-Dichloro-β-[(p-chlorobenzyl)oxy}phenethyl]imidazole mononitrate
Empirical Formula (Hill Notation):
C18H16Cl3N3O4
CAS Number:
Molecular Weight:
444.70
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing